missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Octene, 97%, Thermo Scientific Chemicals

Product Code. 11304838
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11304838

Brand: Thermo Scientific Alfa Aesar 043154.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14919-01-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00039971
InChI Key YCTDZYMMFQCTEO-FNORWQNLSA-N
Synonym trans-3-octene, e-3-octene, e-oct-3-ene, 3-octene, e, 3-octene, 3e, unii-2a4p9ko860, 3-octene, 3e-3-octene, 3e-oct-3-ene, cis-3-octene
PubChem CID 638228
IUPAC Name (E)-oct-3-ene
SMILES CCCCC=CCC

Specifications

Density 0.716 g/mL
Boiling Point 121°C to 222°C
Flash Point 13°C (57°F)
Odor Characteristic
Refractive Index 1.413
Quantity 10 g
UN Number UN3295
Formula Weight 112.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-3-Octene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.