missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-(4-n-Heptylcyclohexyl)benzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as intermediates of liquid crystals.
Solubility
Slightly soluble in water.
Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
It is used as intermediates of liquid crystals.
Solubility
Slightly soluble in water.
Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 33°C |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 283.46 |
Percent Purity | 97% |
Chemical Name or Material | trans-4-(4-n-Heptylcyclohexyl)benzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'