missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(4-n-Pentylcyclohexyl)benzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 15473838
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It has been used for intermediate of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used for intermediate of liquid crystals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65355-30-8
Molecular Formula C18H26O2
Molecular Weight (g/mol) 274.404
MDL Number MFCD06208360
InChI Key YXKKMVGGPRVHIL-UHFFFAOYSA-N
Synonym 4-4-pentylcyclohexyl benzoic acid, 4-trans-4-n-pentylcyclohexyl benzoic acid, 4-trans-4-pentylcyclohexyl benzoic acid, 4-1s,4r-4-pentylcyclohexyl benzoic acid, 4-pentyl-cyclohexyl benzoic acid, 4-pentyl cyclohexyl benzoic acid, yxkkmvggprvhil-shtzxodssa-n, 4-4-pentyl-cyclohexyl-benzoic acid, 4-trans-4-amylcyclohexyl benzoic acid
PubChem CID 3104716
IUPAC Name 4-(4-pentylcyclohexyl)benzoic acid
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 274.4
Percent Purity 99%
Chemical Name or Material trans-4-(4-n-Pentylcyclohexyl)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.