missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Aminocyclohexanol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 110°C to 113°C |
Quantity | 100 g |
UN Number | UN3259 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 115.18 |
Percent Purity | ≥98% |
Chemical Name or Material | trans-4-Aminocyclohexanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'