missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanol hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11446057
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50910-54-8
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.63
MDL Number MFCD00012566,MFCD07366531,MFCD06410647
InChI Key RKTQEVMZBCBOSB-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl
PubChem CID 522619
IUPAC Name 4-aminocyclohexan-1-ol;hydrochloride
SMILES Cl.NC1CCC(O)CC1

Specifications

Melting Point 225°C to 230°C
Quantity 100 g
Beilstein 3909294
Sensitivity Hygroscopic
Formula Weight 151.64
Percent Purity 97%
Chemical Name or Material trans-4-Aminocyclohexanol hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'