missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals trans-4-Hydroxy-L-proline, 99+%

Product Code. 11926601
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PubChem CID 5810
ChEBI CHEBI:18095
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)O

Specifications

Melting Point 274.0°C to 275.0°C
Color White
Loss on Drying 0.2% (105°C, 3 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 500 g
Beilstein 22,191
Merck Index 15,4878
Specific Rotation Condition - 75.50 (20.00°C c=5,H2O)
Specific Rotation − 75.50
Solubility Information Solubility in water: 357.8g/L (20° c). Other solubilities: slightly soluble in alcohol, insoluble in ether
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 131.13
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-4-Hydroxy-L-proline
Show More Show Less
Product Identifier
  • trans-4-Hydroxy-L-proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.