missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Hydroxy-L-proline, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Description
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 270°C to 273°C (decomposition) |
Beilstein | 471933 |
Merck Index | 14,4840 |
Solubility Information | Soluble in water. |
Formula Weight | 131.13 |
Percent Purity | ≥99% |
Quantity | 100 g |
Optical Rotation | −76° (c=4 in water) |
Chemical Name or Material | trans-4-Hydroxy-L-proline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'