missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-L-proline, 99+%, Thermo Scientific Chemicals

Catalog Number p-7081745
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
51-35-4
C5H9NO3
131.131
MFCD00064320
PMMYEEVYMWASQN-DMTCNVIQSA-N
trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
5810
CHEBI:18095
(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
C1C(CNC1C(=O)O)O
This item is not returnable. View return policy
51-35-4
C5H9NO3
131.131
MFCD00064320
PMMYEEVYMWASQN-DMTCNVIQSA-N
trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
5810
CHEBI:18095
(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
C1C(CNC1C(=O)O)O

 

trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 270°C to 273°C (decomposition)
Beilstein 471933
Merck Index 14,4840
Solubility Information Soluble in water.
Formula Weight 131.13
Percent Purity ≥99%
Quantity 10 g
Optical Rotation −76° (c=4 in water)
Chemical Name or Material trans-4-Hydroxy-L-proline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'