missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Hydroxycrotonic acid, Tocris Bioscience™
GHB receptor ligand
Chemical Identifiers
CAS | 24587-49-3 |
---|---|
Molecular Formula | C4H6O3 |
Molecular Weight (g/mol) | 102.089 |
InChI Key | RMQJECWPWQIIPW-OWOJBTEDSA-N |
Synonym | 4-hydroxycrotonic acid, trans-4-hydroxycrotonic acid, e-4-hydroxybut-2-enoic acid, 4-hydroxy-2-butenoic acid, e-4-hydroxycrotonic acid, 4-hydroxy-crotonic acid, gamma-hydroxycrotonic acid, 4-hydroxybut-2-enoic acid, 4-hydroxy-but-2-enoic acid, t-hca |
PubChem CID | 6155526 |
IUPAC Name | (E)-4-hydroxybut-2-enoic acid |
SMILES | C(C=CC(=O)O)O |
Description
Binds to the γ-hydroxybutyric acid (GHB) receptor with higher affinity than GHB itself. May be an endogenous ligand.Chemical Identifiers
24587-49-3 | |
102.089 | |
4-hydroxycrotonic acid, trans-4-hydroxycrotonic acid, e-4-hydroxybut-2-enoic acid, 4-hydroxy-2-butenoic acid, e-4-hydroxycrotonic acid, 4-hydroxy-crotonic acid, gamma-hydroxycrotonic acid, 4-hydroxybut-2-enoic acid, 4-hydroxy-but-2-enoic acid, t-hca | |
(E)-4-hydroxybut-2-enoic acid |
C4H6O3 | |
RMQJECWPWQIIPW-OWOJBTEDSA-N | |
6155526 | |
C(C=CC(=O)O)O |
Specifications
24587-49-3 | |
4-hydroxycrotonic acid, trans-4-hydroxycrotonic acid, e-4-hydroxybut-2-enoic acid, 4-hydroxy-2-butenoic acid, e-4-hydroxycrotonic acid, 4-hydroxy-crotonic acid, gamma-hydroxycrotonic acid, 4-hydroxybut-2-enoic acid, 4-hydroxy-but-2-enoic acid, t-hca | |
C(C=CC(=O)O)O | |
102.089 | |
102.09 | |
trans-4-Hydroxycrotonic acid |
C4H6O3 | |
RMQJECWPWQIIPW-OWOJBTEDSA-N | |
(E)-4-hydroxybut-2-enoic acid | |
6155526 | |
>99% |
Safety and Handling
Recommended Storage : Desiccate at +4°C