missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Methoxycinnamaldehyde, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with strong oxidizing agents.
trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 56°C to 60°C |
Boiling Point | 160°C (3 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Pleasant |
Quantity | 50 g |
Beilstein | 1447540 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 162.19 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'