Learn More
trans-4-Methoxycinnamic acid, 98%
Description
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.
Solubility
Soluble in ethanol, ethyl acetate and other organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 171°C to 175°C |
| Quantity | 50 g |
| Beilstein | 510468 |
| Solubility Information | Soluble in ethanol,ethyl acetate and other organic solvents. |
| Formula Weight | 178.19 |
| Percent Purity | 98% |
| Chemical Name or Material | trans-4-Methoxycinnamic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.