missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methylcyclohexanol, 98%, Thermo Scientific Chemicals

£98.40 - £328.00

Chemical Identifiers

CAS 7731-29-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00064171
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(CC1)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11440531
View Documents
Thermo Scientific Alfa Aesar
B20054.06
5 g
£98.40
5g
Please to purchase this item. Need a web account? Register with us today!
11450531
View Documents
Thermo Scientific Alfa Aesar
B20054.14
25 g
£328.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

7731-29-5
114.188
MQWCXKGKQLNYQG-UHFFFAOYSA-N
11524
CC1CCC(CC1)O
C7H14O
MFCD00064171
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
4-methylcyclohexan-1-ol

Specifications

7731-29-5
173°C to 175°C
C7H14O
MFCD00064171
Slightly soluble in water.
CC1CCC(CC1)O
114.188
114.19
trans-4-Methylcyclohexanol
0.91
72°C (162°F)
1.455
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
MQWCXKGKQLNYQG-UHFFFAOYSA-N
4-methylcyclohexan-1-ol
11524
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 231-790-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only