missing translation for 'onlineSavingsMsg'
Learn More

trans-4-tert-Pentylcyclohexanol, 97%, Thermo Scientific™

Product Code. 11437014
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20698-30-0
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00067157,MFCD00019356
InChI Key VDQZABQVXYELSI-UHFFFAOYSA-N
Synonym 4-tert-amylcyclohexanol, 4-tert-pentyl cyclohexanol, trans-4-tert-pentyl cyclohexanol, p-tert-amylcyclohexanol, trans-4-tert-pentylcyclohexanol, cyclohexanol, 4-tert-pentyl, cyclohexanol, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl cyclohexan-1-ol, 4-tert-pentylcyclohexanol, 4-2-methylbutan-2-yl cyclohexanol
PubChem CID 79302
IUPAC Name 4-(2-methylbutan-2-yl)cyclohexan-1-ol
SMILES CCC(C)(C)C1CCC(O)CC1

Specifications

Melting Point 24°C to 27°C
Boiling Point 154°C to 156°C (40 mmHg)
Quantity 5 g
Formula Weight 170.3
Percent Purity 97%
Chemical Name or Material trans-4-tert-Pentylcyclohexanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'