Learn More
Chemical Identifiers
CAS | 4180-23-8 |
---|---|
Molecular Formula | C10H12O |
Molecular Weight (g/mol) | 148.205 |
MDL Number | MFCD00009284 |
InChI Key | RUVINXPYWBROJD-ONEGZZNKSA-N |
Synonym | anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole |
PubChem CID | 637563 |
ChEBI | CHEBI:35616 |
IUPAC Name | 1-methoxy-4-[(E)-prop-1-enyl]benzene |
SMILES | CC=CC1=CC=C(C=C1)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11489040
|
Thermo Scientific Alfa Aesar
A13482.14 |
25 g |
£20.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499040
|
Thermo Scientific Alfa Aesar
A13482.22 |
100 g |
£34.47
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409050
|
Thermo Scientific Alfa Aesar
A13482.36 |
500 g |
£111.56
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.
Solubility
Soluble in benzene, ethyl acetate, acetone, carbon disulfide.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
4180-23-8 | |
148.205 | |
RUVINXPYWBROJD-ONEGZZNKSA-N | |
637563 | |
1-methoxy-4-[(E)-prop-1-enyl]benzene |
C10H12O | |
MFCD00009284 | |
anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole | |
CHEBI:35616 | |
CC=CC1=CC=C(C=C1)OC |
Specifications
4180-23-8 | |
0.991 | |
96°C (205°F) | |
1.56 | |
774229 | |
14,643 | |
Soluble in benzene,ethyl acetate,acetone,carbon disulfide. | |
CC=CC1=CC=C(C=C1)OC | |
148.205 | |
CHEBI:35616 | |
≥98% |
20°C to 23°C | |
232°C to 235°C | |
C10H12O | |
MFCD00009284 | |
Light sensitive | |
anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole | |
RUVINXPYWBROJD-ONEGZZNKSA-N | |
1-methoxy-4-[(E)-prop-1-enyl]benzene | |
637563 | |
148.21 | |
trans-Anethole, stabilized |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
missing translation for 'einecsNumber' : 224-052-0
missing translation for 'rtecsNumber' : BZ9275000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- trans-Anethole
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : trans-Anethole
RUO – Research Use Only