missing translation for 'onlineSavingsMsg'
Learn More
trans-Anethole, 98+%, Thermo Scientific Chemicals Product Code.: 11489040

trans-Anethole, 98+%, Thermo Scientific Chemicals

Product Code. 11489040
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11489040

Brand: Thermo Scientific Alfa Aesar A13482.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.

Solubility
Soluble in benzene, ethyl acetate, acetone, carbon disulfide.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4180-23-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009284
InChI Key RUVINXPYWBROJD-ONEGZZNKSA-N
Synonym anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole
PubChem CID 637563
ChEBI CHEBI:35616
IUPAC Name 1-methoxy-4-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=C(C=C1)OC

Specifications

Melting Point 20°C to 23°C
Density 0.991
Boiling Point 232°C to 235°C
Flash Point 96°C (205°F)
Refractive Index 1.56
Quantity 25 g
Beilstein 774229
Sensitivity Light sensitive
Merck Index 14,643
Solubility Information Soluble in benzene,ethyl acetate,acetone,carbon disulfide.
Formula Weight 148.21
Percent Purity ≥98%
Chemical Name or Material trans-Anethole, stabilized
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252187 Certificate TypeCertificate of Analysis Date26/08/2024 Catalog NumberA13482.22,A13482.14,A13482.36
Lot Number10250316 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberA13482.22,A13482.14,A13482.36
Lot Number10220697 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog NumberA13482.22,A13482.14,A13482.36
Lot Number10235591 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13482.22,A13482.14,A13482.36
Lot Number10195972 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13482.22,A13482.14,A13482.36
compliance-icons
Product Identifier
  • trans-Anethole
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : trans-Anethole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-Anethole, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.