missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Bis(benzonitrile)dichloropalladium(II), Pd 27.1%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Linear Formula | PdCl2[(C6H5)(CN)]2 |
Quantity | 0.5 g |
UN Number | UN3439 |
Solubility Information | Soluble in acetone, chloroform |
Formula Weight | 383.57 |
Concentration or Composition (by Analyte or Components) | Pd 27.1% |
Physical Form | Powder |
Chemical Name or Material | trans-Bis(benzonitrile)dichloropalladium(II) |
Safety and Handling
Product Identifier
- trans-Bis(benzonitrile)dichloropalladium(II)
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'