missing translation for 'onlineSavingsMsg'
Learn More

trans-Chalcone, 97%

Product Code. 11457776
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11457776

Brand: Thermo Scientific Alfa Aesar A14734.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-47-1 | C15H12O | 208.26 g/mol

Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.

Solubility
Soluble in chloroform, ether, benzene, and ethanol (slightly). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-47-1
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003082
InChI Key DQFBYFPFKXHELB-VAWYXSNFSA-N
Synonym chalcone, trans-chalcone, e-chalcone, benzalacetophenone, chalkone, benzylideneacetophenone, phenyl styryl ketone, cinnamophenone, 2-benzalacetophenone, 2-benzylideneacetophenone
PubChem CID 637760
ChEBI CHEBI:48965
IUPAC Name (E)-1,3-diphenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 55°C to 57°C
Boiling Point 208°C (25 mmHg)
Quantity 500 g
Beilstein 509985
Merck Index 14,2037
Solubility Information Soluble in chloroform,ether,benzene,and ethanol (slightly). Insoluble in water.
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material trans-Chalcone
compliance-icons
Product Identifier
  • trans-Chalcone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : (E)-Chalcone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.