missing translation for 'onlineSavingsMsg'
Learn More
trans-Cinnamaldehyde, 98+%, Thermo Scientific Chemicals Product Code.: 11457686

trans-Cinnamaldehyde, 98+%, Thermo Scientific Chemicals

Product Code. 11457686
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11457686

Brand: Thermo Scientific Alfa Aesar A14689.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

trans-Cinnamaldehyde is used in the flavor and perfume industry. It is also used in medicine. It reacts with glutathione to get an adduct 1'-(glutathion-S-yl)-dihydrocinnamaldehyde. It is used to prepare cinnamylidene-bisacetamide by reacting with acetamide. Further, it inhibits xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Cinnamaldehyde is used in the flavor and perfume industry. It is also used in medicine. It reacts with glutathione to get an adduct 1′-(glutathion-S-yl)-dihydrocinnamaldehyde. It is used to prepare cinnamylidene-bisacetamide by reacting with acetamide. Further, it inhibits xanthine oxidase.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14371-10-9
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00007000
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
SMILES O=C\C=C\C1=CC=CC=C1

Specifications

Melting Point °C to 8°C
Density 1.050
Boiling Point 250°C to 252°C
Flash Point 131°C (268°F)
Assay Percent Range ≥98%
Refractive Index 1.622
Quantity 500 g
Beilstein 1071571
Sensitivity Air Sensitive
Merck Index 14,2297
Solubility Information Miscible with water.
IUPAC Name (2E)-3-phenylprop-2-enal
Formula Weight 132.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material trans-Cinnamaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456557 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog NumberA14689.0E,A14689.22,A14689.36
Lot NumberA0444820 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog NumberA14689.0E,A14689.22,A14689.36
Lot Number10243951 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA14689.0E,A14689.22,A14689.36
Lot Number10243456 Certificate TypeCertificate of Analysis Date13/09/2022 Catalog NumberA14689.0E,A14689.22,A14689.36
Lot Number10243453 Certificate TypeCertificate of Analysis Date13/09/2022 Catalog NumberA14689.0E,A14689.22,A14689.36
compliance-icons
Product Identifier
  • trans-Cinnamaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : trans-Cinnamaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-Cinnamaldehyde, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.