Learn More
trans-Cinnamaldehyde, 98+%, Thermo Scientific Chemicals
Description
trans-Cinnamaldehyde is used in the flavor and perfume industry. It is also used in medicine. It reacts with glutathione to get an adduct 1'-(glutathion-S-yl)-dihydrocinnamaldehyde. It is used to prepare cinnamylidene-bisacetamide by reacting with acetamide. Further, it inhibits xanthine oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-Cinnamaldehyde is used in the flavor and perfume industry. It is also used in medicine. It reacts with glutathione to get an adduct 1′-(glutathion-S-yl)-dihydrocinnamaldehyde. It is used to prepare cinnamylidene-bisacetamide by reacting with acetamide. Further, it inhibits xanthine oxidase.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases. Air sensitive.
Specifications
Specifications
Melting Point | -8°C |
Density | 1.05 |
Boiling Point | 250°C to 252°C |
Flash Point | 131°C (268°F) |
Assay Percent Range | ≥98% |
Odor | Characteristic |
Refractive Index | 1.622 |
Quantity | 2500 g |
Beilstein | 1071571 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.