missing translation for 'onlineSavingsMsg'
Learn More
trans-Cinnamic acid, 99+%, Thermo Scientific Chemicals Product Code.: 11480004

trans-Cinnamic acid, 99+%, Thermo Scientific Chemicals

Product Code. 11480004
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11480004

Brand: Thermo Scientific Alfa Aesar A13538.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

trans-Cinnamic acid is used in flavors, synthetic indigo and pharmaceuticals. It is involved in the production of methyl, ethyl and benzyl esters, which is used in the perfume industry. It serves as a precursor to the sweetener aspartame through enzyme-catalyzed amination to phenylalanine. It is a self-inhibitor produced by fungal spores to prevent germination. In addition, it is used to establish phenolic compounds by liquid chromatography, ultraviolet and mass spectrometry. It is utilized as a potential agent, thereby preventing lung tumor cells from metastasizing. Further, it induces intracellular release of calcium ions from the vacuole to the cytoplasm in order to trigger phytotoxicity in cucumber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Cinnamic acid is used in flavors, synthetic indigo and pharmaceuticals. It is involved in the production of methyl, ethyl and benzyl esters, which is used in the perfume industry. It serves as a precursor to the sweetener aspartame through enzyme-catalyzed amination to phenylalanine. It is a self-inhibitor produced by fungal spores to prevent germination. In addition, it is used to establish phenolic compounds by liquid chromatography, ultraviolet and mass spectrometry. It is utilized as a potential agent, thereby preventing lung tumor cells from metastasizing. Further, it induces intracellular release of calcium ions from the vacuole to the cytoplasm in order to trigger phytotoxicity in cucumber.

Solubility
Soluble in many organic solvents. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-10-3
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00004369
InChI Key WBYWAXJHAXSJNI-VOTSOKGWSA-N
Synonym cinnamic acid, trans-cinnamic acid, e-cinnamic acid, 3-phenylacrylic acid, trans-3-phenylacrylic acid, zimtsaeure, 2e-3-phenylprop-2-enoic acid, 3-phenylprop-2-enoic acid, 3-phenylpropenoic acidShow MoreShow Less
PubChem CID 444539
ChEBI CHEBI:35697
IUPAC Name (E)-3-phenylprop-2-enoic acid
SMILES C1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 133°C to 136°C
Boiling Point 147°C (4 mm)
Assay Percent Range ≥99%
Beilstein 1905952
Merck Index 14,2299
Solubility Information Soluble in many organic solvents. Slightly soluble in water.
Formula Weight 148.16
Percent Purity ≥99%
Density 0.910
Quantity 100 g
Flash Point 166°C (330°F)
Chemical Name or Material trans-Cinnamic Acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW13K012 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog NumberA13538.22,A13538.36,A13538.0E
Lot NumberS13K060 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog NumberA13538.22,A13538.36,A13538.0E
Lot NumberQ16K005 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA13538.22,A13538.36,A13538.0E
Lot NumberN19K009 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberA13538.22,A13538.36,A13538.0E
Lot NumberW06J010 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog NumberA13538.22,A13538.36,A13538.0E
compliance-icons
Product Identifier
  • trans-Cinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : trans-Cinnamic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-Cinnamic acid, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.