missing translation for 'onlineSavingsMsg'
Learn More

trans-Diamminedichloroplatinum(II), Pt 64.5% min, Thermo Scientific Chemicals

£150.75 - £588.93

Chemical Identifiers

CAS 14913-33-8
Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.05
MDL Number MFCD00011623
InChI Key LXZZYRPGZAFOLE-UHFFFAOYSA-L
Synonym trans-dichlorodiamineplatinum ii, azane; dichloroplatinum, h6cl2n2pt, ia call, ddp-h, platinol tn, cl.2h3n.pt, platinum ii chloride diamine, spectrum1502107, cisplatin jp17/usp/inn
PubChem CID 5702198
ChEBI CHEBI:27899
IUPAC Name azane;dichloroplatinum
SMILES N.N.Cl[Pt]Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355978
View Documents
Thermo Scientific Alfa Aesar
010472.03
1 g
£150.75
1g
Please to purchase this item. Need a web account? Register with us today!
11365978
View Documents
Thermo Scientific Alfa Aesar
010472.06
5 g
£588.93
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

trans-Diamminedichloroplatinum(II) acts as a reversible RNA-protein cross-linking agent. It is also used in plating baths. It reacts with DNA after hydrolysis to get both intra and interstrand crosslinks. These crosslinks appear to impair replication and transcription of DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14913-33-8
300.05
LXZZYRPGZAFOLE-UHFFFAOYSA-L
5702198
azane;dichloroplatinum
Cl2H6N2Pt
MFCD00011623
trans-dichlorodiamineplatinum ii, azane; dichloroplatinum, h6cl2n2pt, ia call, ddp-h, platinol tn, cl.2h3n.pt, platinum ii chloride diamine, spectrum1502107, cisplatin jp17/usp/inn
CHEBI:27899
N.N.Cl[Pt]Cl

Specifications

340°C (dec.)
Yellow
Cl2H6N2Pt
MFCD00011623
Soluble in dimethylsulfoxide and dimethylformamide. Slightly soluble in water.
N.N.Cl[Pt]Cl
300.05
CHEBI:27899
Pt 64.5% min.
trans-Diamminedichloroplatinum(II)
14913-33-8
Crystalline
PtCl2(NH3)2
trans-dichlorodiamineplatinum ii, azane; dichloroplatinum, h6cl2n2pt, ia call, ddp-h, platinol tn, cl.2h3n.pt, platinum ii chloride diamine, spectrum1502107, cisplatin jp17/usp/inn
LXZZYRPGZAFOLE-UHFFFAOYSA-L
azane;dichloroplatinum
5702198
300.06
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H341-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H317-H318-H334-H340-H351

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 238-980-9

missing translation for 'rtecsNumber' : TP2455000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only