Learn More
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min, Thermo Scientific Chemicals
Description
As a catalyst of choice in hydrogenation, hydrosilation, carbonylation, oxidation, elimination, reduction, allyl aceate isomerization, dimerization of 1,3-dienes, and C-C bond formation such as Negishi coupling, Suzuki coupling, Kumada coupling, Sonogashira coupling, Heck coupling and Sonogashira-Hagihara coupling reactions. Used in the preparation of tetrakis(triphenylphosphine)palladium, condensed heterocycles such as furoquinolines, diphenylacetylenes, and several other compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼310°C (decomposition) |
Color | Yellow |
Odor | Odorless |
Linear Formula | PdCl2[P(C6H5)3]2 |
Quantity | 25 g |
Sensitivity | Hygroscopic |
Solubility Information | Insoluble in water. Soluble in benzene,and toluene. |
Formula Weight | 701.91 |
Concentration or Composition (by Analyte or Components) | Pd 14.0% min. |
Physical Form | Crystalline |
Show More |
RUO – Research Use Only