missing translation for 'onlineSavingsMsg'
Learn More
trans-N-Boc-4-[2-(trifluoromethyl)benzyl]-L-proline, 95%, Thermo Scientific Chemicals Product Code.: 15419625

trans-N-Boc-4-[2-(trifluoromethyl)benzyl]-L-proline, 95%, Thermo Scientific Chemicals

Product Code. 15419625
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15419625

Brand: Thermo Scientific Alfa Aesar H52137.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957311-13-6
Molecular Formula C18H22F3NO4
Molecular Weight (g/mol) 373.372
MDL Number MFCD16658835
InChI Key LXKNKSPHSOFZBM-RISCZKNCSA-N
Synonym 2s,4r-1-tert-butoxycarbonyl-4-2-trifluoromethyl benzyl pyrrolidine-2-carboxylic acid, 2s,4r-1-tert-butoxycarbonyl-4-2-trifluoromethyl phenyl methyl pyrrolidine-2-carboxylic acid, boc-r-gamma-2-trifluoromethyl-benzyl-l-proline, boc-r-4-2-trifluoromethyl benzyl-l-proline, trans-n-boc-4-2-trifluoromethyl benzyl-l-proline, boc-r-4-2-trifluoromethyl benzyl-l-proline hplc, 2s,4r-1-tert-butoxycarbonyl-4-2-trifluoromethyl benzyl pyrrolidine-2-carboxylicacid, 4r-1-tert-butoxycarbonyl-4-2-trifluoromethyl phenyl methyl-l-proline, 2s,4r-1-tert-butoxycarbonyl-4-2-trifluoromethyl-benzyl pyrrolidine-2-carboxylic acid, 2s,4r-1-2-methylpropan-2-yl oxycarbonyl-4-2-trifluoromethyl phenyl methyl pyrrolidine-2-carboxylic acid
PubChem CID 2762026
IUPAC Name (2S,4R)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-[[2-(trifluoromethyl)phenyl]methyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)CC2=CC=CC=C2C(F)(F)F

Specifications

Melting Point 117°C to 119°C
Quantity 1 g
Formula Weight 373.37
Percent Purity 95%
Chemical Name or Material trans-N-Boc-4-[2-(trifluoromethyl)benzyl]-L-proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-N-Boc-4-[2-(trifluoromethyl)benzyl]-L-proline, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.