missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Decadienal, 95%, Tech., Thermo Scientific Chemicals

Product Code. 10502781
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25152-84-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00007007
InChI Key JZQKTMZYLHNFPL-BLHCBFLLSA-N
Synonym 2e,4e-deca-2,4-dienal, 2,4-decadienal, e,e-2,4-decadienal, trans,trans-2,4-decadienal, 2,4-decadienal, 2e,4e, 2e,4e-decadienal, 2e,4e-2,4-decadienal, 2,4-decadienal, e,e, 2-trans-4-trans-decadienal, ccris 4029
PubChem CID 5283349
IUPAC Name (2E,4E)-deca-2,4-dienal
SMILES CCCCCC=CC=CC=O

Specifications

Color Yellow
Density 0.8500g/mL
Boiling Point 114°C to 116°C (10.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (main isomer) (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH=CHCH=CHCHO
Refractive Index 1.5140 to 1.518
Quantity 5 g
Specific Gravity 0.85
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 152.24
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material trans, trans-2, 4-Decadienal
Show More Show Less
compliance-icons
Product Identifier
  • trans,trans-2,4-Decadienal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'