missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-2,4-Hexadienyl acetate, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
trans,trans-2,4-Hexadienyl acetate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
trans,trans-2,4-Hexadienyl acetate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
Density | 0.93 |
Boiling Point | 100°C (20 mmHg) |
Flash Point | 71°C (159°F) |
Refractive Index | 1.4767 |
Quantity | 5 g |
Beilstein | 1702607 |
Solubility Information | Insoluble in water. |
Formula Weight | 140.18 |
Percent Purity | 97% |
Chemical Name or Material | trans,trans-2,4-Hexadienyl acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'