missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin, 97%, Thermo Scientific Chemicals

Plant growth hormone that stimulates cell division

£219.09 - £782.90

Chemical Identifiers

CAS 1637-39-4
Molecular Formula C10H13N5O
Molecular Weight (g/mol) 219.248
MDL Number MFCD00213654
InChI Key UZKQTCBAMSWPJD-FARCUNLSSA-N
Synonym trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e
PubChem CID 449093
ChEBI CHEBI:16522
IUPAC Name (E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
SMILES CC(=CCNC1=NC=NC2=C1NC=N2)CO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15494449
View Documents
Thermo Scientific Alfa Aesar
J61972.MF
50 mg
£219.09
50mg
Please to purchase this item. Need a web account? Register with us today!
15474449
View Documents
Thermo Scientific Alfa Aesar
J61972.MC
100 mg
£327.63
100mg
Please to purchase this item. Need a web account? Register with us today!
15484449
View Documents
Thermo Scientific Alfa Aesar
J61972.MD
250 mg
£782.90
250mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.

Solubility
Soluble in DMSO and DMF

Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
Specifications

Chemical Identifiers

1637-39-4
219.248
UZKQTCBAMSWPJD-FARCUNLSSA-N
449093
(E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
C10H13N5O
MFCD00213654
trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e
CHEBI:16522
CC(=CCNC1=NC=NC2=C1NC=N2)CO

Specifications

1637-39-4
White
MFCD00213654
14,10117
Soluble in DMSO and DMF
CC(=CCNC1=NC=NC2=C1NC=N2)CO
219.248
CHEBI:16522
97%
trans-Zeatin
208°C to 210°C
C10H13N5O
616241
trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e
UZKQTCBAMSWPJD-FARCUNLSSA-N
(E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
449093
219.25
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : EM9506000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only