missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Transferpette™ -12 Electronic Microliter Pipette

Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.

Brand:  Brand™ 705460

956.44 GBP valid until 2025-12-31
Use promo code "24313" to get your promotional price.


Product Code. 12634060

  • £1095.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Functional, ergonomic housing design
  • 12 channels
  • Individually adjustable finger rest
  • Intuitive menu structure and comprehensively illustrated user manual
  • Significantly reduced tip attachment and ejection forces using universal tip
  • Corrosion-resistant piston and ejector
  • Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis, dispensing
  • Ready for use: 4000 pipetting cycles with each battery charge, battery refresh function even during recharging
  • Each pipette has a DE-M marking and is supplied with performance certificate
  • Supplied with battery, AC adapter, device stand, reagent reservoir, mounting tool, silicone oil and one set of sealing rings made of FKM

TRUSTED_SUSTAINABILITY
Specifications

Specifications

≤ 1.0%, ± 0.2μL
Autoclavable
12
Yes
1 to 20 μL
Ergonomic
0.02μL
230V/50 Hz
360°
Gray
Transferpette™
Electronic Microliter Pipette
LCD
UK/Ireland Adapter
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.