missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Multi-Channel Micropipette Transferpette™ -8 Electronic, DE-M

Product Code. 12398603 Shop All Brand Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 20 μL
10 to 200 μL
15 to 300 μL
5 to 100 μL
Color:
Gray
Green
Yellow
Includes:
UK/Ireland AC Adapter
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12398603

Brand: BRAND™ 705410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.

  • Transferpette electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
  • Features: The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
  • Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μl also GEL-Electrophoresis.
    Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
  • Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ≤ 1.0%, ± 0.2μL
Angle 360°
Autoclavable Autoclavable
Color Gray
Number of Channels 8
Product Line Transferpette
Type Multi-Channel Pipette
Volume (Metric) 1 to 20 μL
Display LCD
Finger Style Ergonomic
Includes UK/Ireland AC Adapter
Increments 0.02μL
Quantity 1 Each
Electrical Requirements 230V/50 Hz
Voltage 230 V
Frequency 50 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.