missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Single-Channel Micropipette Transferpette™ S, Fixed-Volume, DE-M
GREENER_CHOICE

Efficient: one-hand operation. Flexible: ergonomic design for all hand sizes.

Brand:  BRAND™ 705808

121.29 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 16477785

  • ÂŁ171.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Transferpette™ S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette™ S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip.

  • The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment.
  • The Transferpette™ S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools.
  • A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
  • Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIN EN 285
Autoclavable
Non-sterile
10 ÎĽL
36 Months
28.7 cm
12 cm
4.8 cm
Transferpette™ S, 10 μL, fixed volume, F-10, DE-M
Polypropylene
Fixed Volume Pipette
15 to 40°C
11.3 in.
4.7 in.
1.9 in.
292.80 g
Product Suggestions

Product Suggestions

Videos

Videos

  • Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Brand Transferpette S (EN)

    SDS
    Documents Certificates
    View accessory products
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    BRAND™ Single-Channel Micropipette Transferpette™ S, Fixed-Volume, DE-M > Volume: 10 ÎĽL

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.