Learn More
BRAND™ Transferpette™ S Pipette Shelf Mount
Accessory for use with Transferpette™ S single and multichannel pipettes.
Brand:  BRAND™ 704811
Additional Details : Weight : 0.01480kg
Product Code. 16425999
  • £13.30 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Allows hanging of pipette on shelf, or as a replacement part for benchtop rack
  • Pipettes sold separately
Specifications
Specifications
Shelf/Rack Mount
Transferpette™ S Pipettes