missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Translucent Polypropylene Y-shape Tubing Connector
Constructed of translucent polypropylene. Accommodates three tubes with Y-shaped stem and arms.
Brand: BRAND™ 152900
19.55 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Translucent polypropylene construction
- Accommodates 3 tubes of the same diameter
Specifications
Tubing Connector | |
24 mm | |
20 Pack | |
Y | |
Translucent |
Polypropylene | |
3 mm tubing I.D. | |
3 mm tubing I.D. | |
14 mm distance b/w two arms |