missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Translucent Yellow Viton 3-stopped Pump Tubing
Brand: Masterflex™ 97629-48
186.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
2.79 mm | |
almost any pumping application | |
0.59 to 89mL/min. | |
22 psi |
Viton | |
-50°C | |
Translucent Yellow | |
74°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ Translucent Yellow Viton 3-stopped Pump Tubing > DiameterMetricInside
Spot an opportunity for improvement?Share a Content Correction