missing translation for 'onlineSavingsMsg'
Learn More

Transparent Glass Staining Trough

Prevent evaporation of staining liquid using the drop-on lid supplied with the Transparent Glass Staining Trough. This heavy glass staining jar is an essential microscope accessory for the laboratory.

Brand:  N/A 2956

 View more versions of this product

Product Code. 12868735

  • £64.65 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Glass trough with lid
  • Holds microscope slides 76 x 26mm
  • Trough has ridges inside to hold slides
Specifications

Specifications

Closure Included
Flat
Soda Lime Glass
For microscopic slides
50 mm
Glass
Clear
Staining Trough
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.