missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Narrow Mouth LDPE Safety Wash Bottles

Work safely with potentially dangerous chemicals. VITLAB™ VITsafe™ Narrow Mouth LDPE Safety Wash Bottles feature a permanent safety imprint, color-coded VENT-CAP and precise spray control to eliminate drips and leaks and ensure safe handling.

Brand:  VITLAB™ 1331869

 View more versions of this product

Product Code. 11847900

  • £84.80 / Pack of 12
Estimated Shipment: 09-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

VITsafe™ Narrow Mouth LDPE Safety Wash Bottles – Wash bottles meet all safety requirements for working with potentially dangerous chemicals.
Specifications

Specifications

250 mL
LDPE, Polypropylene
12Pack
GL 25
135 mm
Ethanol
Vented Screw Cap
Transparent
Individually wrapped in PE bag
Orange
Narrow
Safety Wash Bottle
58 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers