Learn More
Fisherbrand™ Non-sterile Transfer Pipettes, Transparent LDPE
Prevent cross contamination. Fisherbrand™ Non-sterile Transfer Pipets, Transparent LDPE are made of shatterproof, flexible plastic and are designed to replace glass Pasteur pipets. Graduated and non-graduated pipets available in a range of sizes and styles, including jumbo, thin stem and extra long.
£7.14 - £35.64
Product Code | Brand | Volume (Metric) | Drops/mL | Graduated | Tip Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Drops/mL | Graduated | Tip Style | Price | Quantity & Availability | ||
13469118
|
Fisherbrand™
|
1 mL | 30 | No | Standard |
£10.12
Pack of 400 |
In Stock
Log in to see stock. |
||
13499108
|
Fisherbrand™
|
1 mL | 30 | Yes | Standard |
£16.38
Pack of 500 |
In Stock
Log in to see stock. |
||
13439118
|
Fisherbrand™
|
1 mL | 30 | Yes | Extended |
£23.55
Pack of 500 |
In Stock
Log in to see stock. |
||
13439108
|
Fisherbrand™
|
3 mL | 25 | Yes | Standard |
£18.06
Pack of 500 |
In Stock
Log in to see stock. |
||
13459118
|
Fisherbrand™
|
4 mL | 30 | No | Thin Stem |
£15.84
Pack of 500 |
In Stock
Log in to see stock. |
||
13459108
|
Fisherbrand™
|
4 mL | 50 | No | Standard |
£35.64
Pack of 500 |
In Stock
Log in to see stock. |
||
13449108
|
Fisherbrand™
|
7 mL | 20 | No | Extra Long |
£7.14
Pack of 100 |
In Stock
Log in to see stock. |
||
13449118
|
Fisherbrand™
|
10 mL | 18 | No | Jumbo |
£7.63
Pack of 200 |
In Stock
Log in to see stock. |
||
Description
- Built-in bulb eliminates the need for plugging, packaging and autoclaving
- Flexible stem bends to sample from narrow or small tubes
- Low affinity surface reduces loss of cells and valuable proteins due to binding
- Soft plastic pipets made of non-toxic low-density polyethylene (LDPE)
- Non-sterile, transparent and fully recyclable
Specifications
LDPE | |
Transfer Pipet |