Learn More
BRAND™ Blaubrand™ Eterna™ AR-GLAS™ Type 3 Class AS Measuring Pipettes
Manufactured from AR-GLAS™ Soda lime glass, this type 3 Class AS Measuring Pipet includes graduations with DE-M marking and is calibrated for total delivery.
£71.40 - £90.60
Specifications
Certifications/Compliance | DIN EN ISO 835 |
---|---|
Material | AR-GLAS |
Graduated | Yes |
Product Code | Brand | Volume (Metric) | Tolerance | Graduations | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Tolerance | Graduations | Price | Quantity & Availability | |||
12005312
|
BRAND™
28405 |
0.5 mL | ±0.006 mL | 0.01 mL |
£90.60
Pack of 12 |
In Stock
Log in to see stock. |
|||
11803980
|
BRAND™
28412 |
5 mL | ±0.03 mL | 0.1 mL |
£83.40
Pack of 12 |
In Stock
Log in to see stock. |
|||
11833920
|
BRAND™
28406 |
1 mL | ±0.007 mL | 0.1 mL |
£71.40
Pack of 12 |
In Stock
Log in to see stock. |
|||
12068359
|
BRAND™
28409 |
2 mL | ±0.010 mL | 0.02 mL |
£71.40
Pack of 12 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
11868331
|
BRAND™
28411 |
5 mL | ±0.030 mL | 0.05 mL |
£85.80
Pack of 12 |
In Stock
Log in to see stock. |
|||
11843920
|
BRAND™
28413 |
10 mL | ±0.05 mL | 0.1 mL |
£85.80
Pack of 12 |
In Stock
Log in to see stock. |
|||
11813980
|
BRAND™
28414 |
20 mL | ±0.10 mL | 0.1 mL |
£87.60
Pack of 6 |
In Stock
Log in to see stock. |
|||
10290021
|
BRAND™
27316 |
1 mL | ±0.007 mL | 0.01 mL |
£87.00
Pack of 12 |
In Stock
Log in to see stock. |
|||
10603561
|
BRAND™
27319 |
2 mL | ±0.070 mL | 0.02 mL |
£87.00
Pack of 12 |
In Stock
Log in to see stock. |
|||
Description
- Manufactured from AR-GLAS™ Soda lime glass
- Type 3 (zero on top)
- DE-M Marking graduations
- Calibrated to deliver (TD, Ex.)
- Supplied with batch number and one batch certificate per packing unit
- USP individual certificate or DAkkS calibration certificate available upon request
Specifications
DIN EN ISO 835 | |
AR-GLAS | |
Yes |