missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Transparent Polypropylene Powder Funnels
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
Brand: Kartell™ 0017000
14.65 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
- 60° angled sides
- External ribbing avoids pressure lock
- Can be used with liquids
- Autoclavable
Specifications
150mm | |
36mm | |
60° | |
Can be used with liquids |
730 mL | |
Polypropylene | |
Autoclavable | |
39mm |