Learn More
Corning™ Transport Medium
Corning™ Transport Medium is an isotonic, neutral pH solution that contains protective proteins, antibiotics to prevent microbial growth and contamination
Brand: Corning™ 25-500-CM
Additional Details : Weight : 5.70000kg
Description
- This isotonic, neutral pH solution contains protective proteins/antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi
- For research and further manufacturing purposes only
- Not for direct use in humans or animals.
- Formulated using the final ingredient concentrations outlined in the CDC's Preparation of Viral Transport Medium (SOP#: DSR-052-01)
- Formulation contains Hanks' Balanced Salt Solution with Ca and Mg (without phenol red), 2% Heat Inactivated Fetal Bovine Serum, 100 μg/mL Gentamicin and 0.5 μg/mL Amphotericin B
Specifications
Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines | |
6/Cs. | |
1 L |
Research, Manufacturing Applications | |
2°C to 8°C |