missing translation for 'onlineSavingsMsg'
Learn More

Transthyretin/Prealbumin Mouse anti-Human, Clone: TTR/4292, Novus Biologicals™

Mouse Monoclonal Antibody

£396.00

Specifications

Antigen Transthyretin/Prealbumin
Clone TTR/4292
Concentration 0.2 mg/mL
Dilution Western Blot 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18635566
View Documents
Novus Biologicals
NBP3-07742-100ug
100 μg
£396.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Transthyretin/Prealbumin Monoclonal specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Transthyretin/Prealbumin
0.2 mg/mL
Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
7276
IgG2c κ
Protein A or G purified
TTR/4292
Western Blot 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Monoclonal
Purified
RUO
Human
ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin
Recombinant human full-length protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.