missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transwab™ Swab with Standing Tube
Click to view available options
:
Swab with Self-standing Tube
Description
Recommended applications:
Features:
- Sampling previously cleaned surfaces
Features:
- Conforms to ISO 18593 and other national and international standards
- Neutralisers allow accurate recovery of residual bacteria on food contact surfaces
- Precise fill for accurate assessment of contamination levels
- 10mL fill
- Reduced packaging & storage space
- Ready to use
- Ambient storage and transport
Specifications
Specifications
| Sterility | Sterile |
| Certifications/Compliance | ISO 18593 |
| Shelf Life | 2 Years |
| Tip Material | Cotton |
| Color | Blue |
| No. per Pack | 50 |
| Cap Color | Blue |
| Quantity | 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction