missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRBP Mouse anti-Human, Clone: 1C9A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66792-1-IG-150UL
This item is not returnable.
View return policy
Description
TRBP is an RNA associated nuclear protein. It contains 3 DRBM (double stranded RNA-binding) domains and plays an important role by binding to HIV-1 TAR RNA located in the long terminal repeat (LTR).TRBP acts synergistic with viral TAT protein in HIV-1 expression activation. It binds preferentially to TAR and other double-stranded RNAs with G+C-rich characteristics. It is important in regulation of transcription from RNA polymerase II promoter andis widely expressed in most of the tissues.Specifications
TRBP | |
Monoclonal | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15633 | |
TARBP2 | |
TRBP Fusion Protein Ag8269 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1C9A10 | |
Unconjugated | |
TARBP2 | |
LOQS, TAR RNA binding protein 2, TARBP2, TRBP, TRBP1, TRBP2 | |
Mouse | |
Protein G | |
RUO | |
6895 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction