Learn More
Uvex Trend BOA™ Black/Blue PU/PU 69382 Low Shoe - Width 11
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Brand:  Uvex 6938241
76.17 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Additional Details : Weight : 1.10500kg
 View more versions of this product
Product Code. 17213911
  • £110.00 / 1 pair
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
Uvex 2 Trend Perforated Shoe with BOA™ Fit System 69382 S1 P SRC
  • Sporty S1 P perforated safety shoe with wide fit
  • Breathable, perforated microvelour
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • BOA™ Fit System Dial for custom comfort that's consistent all day
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole
    • Moisture transport system helps maintain foot temperature
  • Soft padding collar and dust tongue
  • Removable and anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 69382
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 38 to 52
Specifications
Specifications
Black, Blue
41
Steel
EN ISO 20345:2011
Standard Width
Yes
S1
Microvelour
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 2
No
SRC