Learn More
Tri-n-butyl(1-ethoxyvinyl)tin, 95%, Thermo Scientific Chemicals
£45.60 - £560.00
Chemical Identifiers
CAS | 97674-02-7 |
---|---|
Molecular Formula | C16H34OSn |
Molecular Weight (g/mol) | 361.16 |
MDL Number | MFCD00010240 |
InChI Key | HGXJOXHYPGNVNK-UHFFFAOYSA-N |
Synonym | tributyl 1-ethoxyvinyl stannane, tributyl 1-ethoxyvinyl tin, tributyl 1-ethoxyethenyl stannane, 1-ethoxyvinyltributyltin, 1-ethoxyvinyl tributyltin, 1-ethoxyvinyltri-n-butyltin, 1-ethoxyvinyl tributylstannane, 1-ethoxyethenyl tributylstannane, tributyl-1-ethoxyvinyl tin, stannane, tributyl 1-ethoxyethenyl |
PubChem CID | 619414 |
IUPAC Name | tributyl(1-ethoxyethenyl)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)C(=C)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15434157
|
Thermo Scientific Alfa Aesar
H31562.03 |
1 g |
£45.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444157
|
Thermo Scientific Alfa Aesar
H31562.06 |
5 g |
£143.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15454157
|
Thermo Scientific Alfa Aesar
H31562.14 |
25 g |
£560.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97674-02-7 | |
361.16 | |
HGXJOXHYPGNVNK-UHFFFAOYSA-N | |
619414 | |
CCCC[Sn](CCCC)(CCCC)C(=C)OCC |
C16H34OSn | |
MFCD00010240 | |
tributyl 1-ethoxyvinyl stannane, tributyl 1-ethoxyvinyl tin, tributyl 1-ethoxyethenyl stannane, 1-ethoxyvinyltributyltin, 1-ethoxyvinyl tributyltin, 1-ethoxyvinyltri-n-butyltin, 1-ethoxyvinyl tributylstannane, 1-ethoxyethenyl tributylstannane, tributyl-1-ethoxyvinyl tin, stannane, tributyl 1-ethoxyethenyl | |
tributyl(1-ethoxyethenyl)stannane |
Specifications
97674-02-7 | |
85°C to 86°C (0.07 mmHg) | |
C16H34OSn | |
MFCD00010240 | |
tributyl 1-ethoxyvinyl stannane, tributyl 1-ethoxyvinyl tin, tributyl 1-ethoxyethenyl stannane, 1-ethoxyvinyltributyltin, 1-ethoxyvinyl tributyltin, 1-ethoxyvinyltri-n-butyltin, 1-ethoxyvinyl tributylstannane, 1-ethoxyethenyl tributylstannane, tributyl-1-ethoxyvinyl tin, stannane, tributyl 1-ethoxyethenyl | |
CCCC[Sn](CCCC)(CCCC)C(=C)OCC | |
361.16 | |
361.14 |
1.067 | |
>110°C (230°F) | |
1.476 | |
UN2788 | |
HGXJOXHYPGNVNK-UHFFFAOYSA-N | |
tributyl(1-ethoxyethenyl)stannane | |
619414 | |
Tri-n-butyl(1-ethoxyvinyl)tin |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only