missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl(1-propenyl)tin, cis + trans, 94%, Thermo Scientific Chemicals

Product Code. 15489129
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105494-65-3
Molecular Formula C15H32Sn
Molecular Weight (g/mol) 331.131
MDL Number MFCD03425868
InChI Key FJXYMVBFBYAWDR-UHFFFAOYSA-N
Synonym stannane, tributyl-1-propenyl, z-tri-n-butyl 1-propenyl tin, tri-n-butyl 1-propenyl tin, cis + trans, tributyl prop-1-en-1-yl stannane
PubChem CID 57369809
IUPAC Name tributyl(prop-1-enyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=CC

Specifications

Boiling Point 77°C to 78°C (0.25 mmHg)
Quantity 250 mg
UN Number UN2788
Beilstein 4176362
Formula Weight 331.13
Percent Purity 94%
Chemical Name or Material Tri-n-butyl(1-propenyl)tin, Cis + Trans

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.