missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl phosphite, 94%

Product Code. 11363456
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11363456

Brand: Thermo Scientific Alfa Aesar L14307.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-85-2 | C12H27O3P | 250.319 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.

Solubility
Not miscible or difficult to mix in water.

Notes
Air and moisture sensitive. Store under inert gas. Store away from oxidizing agent, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-85-2
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.319
MDL Number MFCD00009437
InChI Key XTTGYFREQJCEML-UHFFFAOYSA-N
Synonym tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri
PubChem CID 7623
IUPAC Name tributyl phosphite
SMILES CCCCOP(OCCCC)OCCCC

Specifications

Melting Point -80°C
Density 0.915
Boiling Point 125°C to 128°C (10 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.432
Quantity 500 mL
Beilstein 1703866
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 250.32
Percent Purity 94%
Chemical Name or Material Tri-n-butyl phosphite
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.