missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin chloride, 95%, tech., Thermo Scientific Chemicals

Product Code. 10684802
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-22-9
Molecular Formula C12H27ClSn
Molecular Weight (g/mol) 325.48
MDL Number MFCD00000521
InChI Key GCTFWCDSFPMHHS-UHFFFAOYSA-M
Synonym tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride
PubChem CID 15096
ChEBI CHEBI:79734
IUPAC Name tributyl(chloro)stannane
SMILES CCCC[Sn](CCCC)(CCCC)Cl

Specifications

Melting Point -9.0°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 171.0°C to 173.0°C (25.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]3SnCl
Refractive Index 1.4880 to 1.4940
Quantity 500 g
Merck Index 15, 9782
Specific Gravity 1.2
Solubility Information Solubility in water: 0.07g/L (20°C). Other solubilities: soluble in most common orgnaic solvents,including,benzene and toluene
Viscosity 7 mPa.s (20°C)
Formula Weight 325.48
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin chloride, tech. 90%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butyltin chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Sicherheitshinweis
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'