missing translation for 'onlineSavingsMsg'
Learn More
Tri-n-butyltin chloride, 95%, tech., Thermo Scientific Chemicals Product Code.: 10755811

Tri-n-butyltin chloride, 95%, tech., Thermo Scientific Chemicals

Product Code. 10755811
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10755811

Brand: Thermo Scientific Acros 139351000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-22-9
Molecular Formula C12H27ClSn
Molecular Weight (g/mol) 325.48
MDL Number MFCD00000521
InChI Key GCTFWCDSFPMHHS-UHFFFAOYSA-M
Synonym tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chlorideShow MoreShow Less
PubChem CID 15096
ChEBI CHEBI:79734
IUPAC Name tributyl(chloro)stannane
SMILES CCCC[Sn](CCCC)(CCCC)Cl

Specifications

Melting Point -9.0°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 171.0°C to 173.0°C (25.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]3SnCl
Refractive Index 1.4880 to 1.4940
Quantity 100 g
Merck Index 15, 9782
Specific Gravity 1.2
Solubility Information Solubility in water: 0.07g/L (20°C). Other solubilities: soluble in most common orgnaic solvents,including,benzene and toluene
Viscosity 7 mPa.s (20°C)
Formula Weight 325.48
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin chloride, tech. 90%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467343 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number139355000,139351000
Lot NumberA0453112 Certificate TypeCertificate of Analysis Date16/05/2023 Catalog Number139355000,139351000
Lot NumberA0449354 Certificate TypeCertificate of Analysis Date12/01/2023 Catalog Number139355000,139351000
Lot NumberA0448701 Certificate TypeCertificate of Analysis Date19/12/2022 Catalog Number
Lot NumberA0399856 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number139355000,139351000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butyltin chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tri-n-butyltin chloride, 95%, tech., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.