missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals

Product Code. 10528883
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-52-3
Molecular Formula C13H30OSn
Molecular Weight (g/mol) 321.07
MDL Number MFCD00009419
InChI Key KJGLZJQPMKQFIK-UHFFFAOYSA-N
Synonym tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl
PubChem CID 16683411
IUPAC Name tributyl(methoxy)stannane
SMILES CCCC[Sn](CCCC)(CCCC)OC

Specifications

Color Colorless
Density 1.1100g/mL
Elemental Analysis 96.0% min.
Boiling Point 97°C to 97.5°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [CH3(CH2)3]3SnOCH3
Refractive Index 1.4726 to 1.4746
Quantity 100 g
Fieser 16,351
Merck Index 15, 9784
Specific Gravity 1.11
Formula Weight 321.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin methoxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butyltin methoxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'