Learn More
Tri-n-octylamine, 95%, Thermo Scientific Chemicals
£50.10 - £695.00
Chemical Identifiers
CAS | 1116-76-3 |
---|---|
Molecular Formula | C24H51N |
Molecular Weight (g/mol) | 353.679 |
MDL Number | MFCD00009560 |
InChI Key | XTAZYLNFDRKIHJ-UHFFFAOYSA-N |
Synonym | trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08 |
PubChem CID | 14227 |
IUPAC Name | N,N-dioctyloctan-1-amine |
SMILES | CCCCCCCCN(CCCCCCCC)CCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11497347
|
Thermo Scientific Alfa Aesar
A15067.AE |
100 mL |
£50.10
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407357
|
Thermo Scientific Alfa Aesar
A15067.AP |
500 mL |
£170.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417357
|
Thermo Scientific Alfa Aesar
A15067.0F |
2500 mL |
£695.00
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTri-n-octylamine is used as an extractant for organic acids like trichloroacetic acid, succinic acid, acetic acid and precious metals. It is also used as a solvent and an intermediate in the manufacture of pharmaceuticals. Further, it is employed in the preparation of quaternary ammonium compounds, agrochemicals, surfactants, lubricant additives, corrosion inhibitors, vulcanization accelerators and dyes.
Solubility
Miscible with chloroform. Immiscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, carbon dioxide, acids, acid chlorides and acid anhydrides.
Chemical Identifiers
1116-76-3 | |
353.679 | |
XTAZYLNFDRKIHJ-UHFFFAOYSA-N | |
14227 | |
CCCCCCCCN(CCCCCCCC)CCCCCCCC |
C24H51N | |
MFCD00009560 | |
trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08 | |
N,N-dioctyloctan-1-amine |
Specifications
1116-76-3 | |
0.81 | |
158°C (316°F) | |
C24H51N | |
1.4485 | |
UN3082 | |
Air Sensitive | |
Miscible with chloroform. Immiscible with water. | |
CCCCCCCCN(CCCCCCCC)CCCCCCCC | |
353.679 | |
353.68 | |
Tri-n-octylamine |
-39°C | |
164°C to 168°C (0.7 mmHg) | |
Amine-like | |
[CH3(CH2)7]3N | |
MFCD00009560 | |
1210618 | |
trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08 | |
XTAZYLNFDRKIHJ-UHFFFAOYSA-N | |
N,N-dioctyloctan-1-amine | |
14227 | |
95% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 214-242-1
missing translation for 'rtecsNumber' : RG8225000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only