missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propyl borate, 98%, Thermo Scientific Chemicals

Product Code. 11352588
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Tri-n-propyl borate is used as solvents or catalysts for the production of resins, waxes, paints and varnishes, antioxidants, flame retardants. It is also used for petroleum products, tackifiers for silicone elastomers, stabilizers and binders for rubbers or plastics and as cross-linking agents in polymerization processes. They are are used in welding fluxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-71-1
Molecular Formula C9H21BO3
Molecular Weight (g/mol) 188.074
MDL Number MFCD00009366
InChI Key LTEHWCSSIHAVOQ-UHFFFAOYSA-N
Synonym tri-n-propyl borate, tripropoxyborane, boron propoxide, propyl orthoborate, tris propoxy borane, boric acid h3bo3 , tripropyl ester, tripropyl orthoborate, boric acid, tripropyl ester, propyl borate, pro 3b, boric acid tri-n-propyl ester
PubChem CID 12711
IUPAC Name tripropyl borate
SMILES B(OCCC)(OCCC)OCCC

Specifications

Color Colorless
Density 0.861g/mL
Boiling Point 175°C to 177°C
Flash Point 32° (90°F)
Linear Formula (C3H7O)3B
Refractive Index 1.3950
Quantity 50 g
UN Number UN3272
Sensitivity Moisture Sensitive
Solubility Information Soluble in organic solvents; Decomposes in water
Formula Weight 188.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tri-n-propyl borate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'